Assay Details
| Gene Symbol: itm2bb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_212976.1 | NP_998141.1 | 4 - 5 | 596 | 81 | |
| GenBank mRNA | BC053278.1 | - | 4 - 5 | 596 | 81 | |
More Information
Set Membership: |
|

itm2bb
Genomic Map