Assay Details
| Gene Symbol: agpat4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_212992.1 | NP_998157.1 | 2 - 3 | 388 | 60 | |
| GenBank mRNA | BC056788.1 | - | 2 - 3 | 388 | 60 | |
| BC155275.1 | - | 2 - 3 | 338 | 60 | ||
More Information
Set Membership: |
|

agpat4
Genomic Map