Assay Details
| Gene Symbol: acbd3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001328207.1 | NP_001315136.1 | 2 - 3 | 469 | 72 | |
| GenBank mRNA | BC054676.1 | - | 2 - 3 | 429 | 72 | |
| BC165020.1 | - | 2 - 3 | 453 | 72 | ||
More Information
Set Membership: |
|

acbd3
Genomic Map