Assay Details
| Gene Symbol: ciao2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_213027.1 | NP_998192.1 | 295 | 73 | ||
| GenBank mRNA | BC059535.1 | - | 295 | 73 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

ciao2a| Gene Symbol: ciao2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_213027.1 | NP_998192.1 | 295 | 73 | ||
| GenBank mRNA | BC059535.1 | - | 295 | 73 | ||
Set Membership: |
|