Assay Details
| Gene Symbol: ciao2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_213027.1 | NP_998192.1 | 1 - 2 | 354 | 61 | |
| GenBank mRNA | BC059535.1 | - | 1 - 2 | 354 | 61 | |
More Information
Set Membership: |
|

ciao2a
Genomic Map