Assay Details
| Gene Symbol: sdf4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_213087.2 | NP_998252.1 | 5 - 6 | 1081 | 79 | |
| GenBank mRNA | BC066581.1 | - | 5 - 6 | 1026 | 79 | |
More Information
Set Membership: |
|

sdf4
Genomic Map