Assay Details
| Gene Symbol: wdr5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_213099.1 | NP_998264.1 | 6 - 7 | 662 | 65 | |
| GenBank mRNA | BC052124.1 | - | 6 - 7 | 716 | 65 | |
| BC066729.1 | - | 6 - 7 | 662 | 65 | ||
More Information
Set Membership: |
|

wdr5
Genomic Map