Assay Details
| Gene Symbol: rpl26 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_213113.2 | NP_998278.1 | 2 - 3 | 212 | 63 | |
| GenBank mRNA | BC055538.1 | - | 2 - 3 | 210 | 63 | |
| BC164945.1 | - | 1 - 2 | 201 | 63 | ||
| EH551244.1 | - | 2 - 3 | 249 | 63 | ||
More Information
Set Membership: |
|

rpl26
Genomic Map