Assay Details
| Gene Symbol: rpl26 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_213113.2 | NP_998278.1 | 3 - 4 | 354 | 56 | |
| GenBank mRNA | BC055538.1 | - | 3 - 4 | 352 | 56 | |
| BC164945.1 | - | 2 - 3 | 343 | 56 | ||
| EH500800.1 | - | 3 - 4 | 376 | 56 | ||
| EH551244.1 | - | 3 - 4 | 391 | 56 | ||
More Information
Set Membership: |
|

rpl26
Genomic Map