Assay Details
| Gene Symbol: dnase1l1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_213153.2 | NP_998318.2 | 4 - 5 | 405 | 94 | |
| GenBank mRNA | BC056304.1 | - | 4 - 5 | 367 | 94 | |
| CN022406.1 | - | 4 - 5 | 478 | 94 | ||
More Information
Set Membership: |
|

dnase1l1
Genomic Map