Assay Details
| Gene Symbol: ap2s1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_213155.2 | NP_998320.1 | 4 - 5 | 483 | 87 | |
| GenBank mRNA | BC058042.1 | - | 425 | 87 | ||
| DT873506.1 | - | 4 - 5 | 487 | 87 | ||
More Information
Set Membership: |
|

ap2s1
Genomic Map