Assay Details
| Gene Symbol: prkag1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_213161.1 | NP_998326.1 | 10 - 11 | 864 | 79 | |
| GenBank mRNA | BC059181.1 | - | 10 - 11 | 864 | 79 | |
More Information
Set Membership: |
|

prkag1
Genomic Map