Assay Details
| Gene Symbol: ca15b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_213182.1 | NP_998347.1 | 5 - 6 | 470 | 83 | |
| XM_005169442.4 | XP_005169499.1 | 5 - 6 | 876 | 83 | ||
| XM_005169443.4 | XP_005169500.1 | 5 - 6 | 821 | 83 | ||
| XM_021480163.1 | XP_021335838.1 | 4 - 5 | 804 | 83 | ||
| GenBank mRNA | BC056289.1 | - | 5 - 6 | 480 | 83 | |
| BC067185.1 | - | 5 - 6 | 470 | 83 | ||
More Information
Set Membership: |
|

ca15b
Genomic Map