Assay Details
| Gene Symbol: ca15b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_213182.1 | NP_998347.1 | 8 - 9 | 772 | 87 | |
| XM_005169442.4 | XP_005169499.1 | 8 - 9 | 87 | |||
| XM_005169443.4 | XP_005169500.1 | 8 - 9 | 87 | |||
| XM_021480163.1 | XP_021335838.1 | 7 - 8 | 87 | |||
| GenBank mRNA | BC056289.1 | - | 8 - 9 | 782 | 87 | |
| BC067185.1 | - | 8 - 9 | 772 | 87 | ||
More Information
Set Membership: |
|

ca15b
Genomic Map