Assay Details
| Gene Symbol: khdrbs1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_213235.1 | NP_998400.1 | 8 - 9 | 1103 | 78 | |
| GenBank mRNA | BC067711.1 | - | 8 - 9 | 1103 | 78 | |
| BC155179.1 | - | 8 - 9 | 1124 | 78 | ||
More Information
Set Membership: |
|

khdrbs1b
Genomic Map