Assay Details
| Gene Symbol: dnm2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_213242.1 | NP_998407.1 | 16 - 17 | 1797 | 61 | |
| GenBank mRNA | BC065325.1 | - | 16 - 17 | 1797 | 61 | |
More Information
Set Membership: |
|

dnm2b
Genomic Map