Assay Details
| Gene Symbol: pabpn1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_213259.2 | NP_998424.1 | 6 - 7 | 1035 | 143 | |
| XM_005162637.3 | XP_005162694.1 | 6 - 7 | 675 | 143 | ||
| GenBank mRNA | BC079522.1 | - | 6 - 7 | 696 | 143 | |
| BC164644.1 | - | 6 - 7 | 692 | 143 | ||
More Information
Set Membership: |
|

pabpn1
Genomic Map