Assay Details
| Gene Symbol: ddah1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_213276.1 | NP_998441.1 | 538 | 72 | ||
| GenBank mRNA | BC068404.1 | - | 538 | 72 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

ddah1| Gene Symbol: ddah1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_213276.1 | NP_998441.1 | 538 | 72 | ||
| GenBank mRNA | BC068404.1 | - | 538 | 72 | ||
Set Membership: |
|