Assay Details
| Gene Symbol: got2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_213379.1 | NP_998544.1 | 8 - 9 | 1077 | 64 | |
| GenBank mRNA | BC054684.1 | - | 8 - 9 | 1077 | 64 | |
| BC165803.1 | - | 8 - 9 | 1048 | 64 | ||
More Information
Set Membership: |
|

got2a
Genomic Map