Assay Details
| Gene Symbol: hnrnpl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_213383.1 | NP_998548.1 | 12 - 13 | 1439 | 116 | |
| XM_005169692.4 | XP_005169749.1 | 12 - 13 | 1583 | 116 | ||
| XM_005169693.4 | XP_005169750.1 | 11 - 12 | 1514 | 116 | ||
| XM_009291459.3 | XP_009289734.1 | 7 - 8 | 1015 | 116 | ||
| GenBank mRNA | BC054655.1 | - | 12 - 13 | 1439 | 116 | |
| BC153481.1 | - | 5 - 6 | 425 | 116 | ||
| BC154328.1 | - | 12 - 13 | 1525 | 116 | ||
More Information
Set Membership: |
|

hnrnpl2
Genomic Map