Assay Details
| Gene Symbol: hnrnpl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_213383.1 | NP_998548.1 | 3 - 4 | 488 | 104 | |
| XM_005169692.4 | XP_005169749.1 | 3 - 4 | 509 | 104 | ||
| XM_005169693.4 | XP_005169750.1 | 3 - 4 | 509 | 104 | ||
| XM_017350996.2 | XP_017206485.1 | 3 - 4 | 509 | 104 | ||
| GenBank mRNA | BC054655.1 | - | 3 - 4 | 488 | 104 | |
| BC154328.1 | - | 3 - 4 | 451 | 104 | ||
| BC165353.1 | - | 3 - 4 | 476 | 104 | ||
More Information
Set Membership: |
|

hnrnpl2
Genomic Map