Assay Details
| Gene Symbol: maf1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_213441.1 | NP_998606.1 | 4 - 5 | 780 | 93 | |
| GenBank mRNA | BC056831.1 | - | 4 - 5 | 780 | 93 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

maf1a| Gene Symbol: maf1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_213441.1 | NP_998606.1 | 4 - 5 | 780 | 93 | |
| GenBank mRNA | BC056831.1 | - | 4 - 5 | 780 | 93 | |
Set Membership: |
|