Assay Details
| Gene Symbol: maf1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_213441.1 | NP_998606.1 | 6 - 7 | 1005 | 74 | |
| GenBank mRNA | BC056831.1 | - | 6 - 7 | 1005 | 74 | |
More Information
Set Membership: |
|

maf1a
Genomic Map