Assay Details
| Gene Symbol: ssbp3b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_213450.1 | NP_998615.1 | 15 - 16 | 1122 | 66 | |
| XM_005166809.4 | XP_005166866.1 | 16 - 17 | 1225 | 66 | ||
| GenBank mRNA | BC056827.1 | - | 15 - 16 | 1122 | 66 | |
More Information
Set Membership: |
|

ssbp3b
Genomic Map