Assay Details
| Gene Symbol: ssbp3b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_213450.1 | NP_998615.1 | 7 - 8 | 589 | 77 | |
| XM_005166809.4 | XP_005166866.1 | 8 - 9 | 692 | 77 | ||
| XM_005166810.4 | XP_005166867.1 | 8 - 9 | 646 | 77 | ||
| XM_009303844.3 | XP_009302119.1 | 7 - 8 | 618 | 77 | ||
| GenBank mRNA | BC056827.1 | - | 7 - 8 | 589 | 77 | |
| GQ903695.1 | - | 8 - 9 | 562 | 77 | ||
More Information
Set Membership: |
|

ssbp3b
Genomic Map