Assay Details
| Gene Symbol: faxdc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_213507.1 | NP_998672.1 | 6 - 7 | 735 | 60 | |
| GenBank mRNA | BC044395.1 | - | 6 - 7 | 735 | 60 | |
More Information
Set Membership: |
|

faxdc2
Genomic Map