Assay Details
| Gene Symbol: p4ha1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_214691.1 | NP_999856.1 | 14 - 15 | 1697 | 104 | |
| XM_009292958.3 | XP_009291233.1 | 14 - 15 | 1789 | 104 | ||
| GenBank mRNA | BC045890.1 | - | 14 - 15 | 1697 | 104 | |
More Information
Set Membership: |
|

p4ha1b
Genomic Map