Assay Details
| Gene Symbol: p4ha1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_214691.1 | NP_999856.1 | 4 - 5 | 477 | 72 | |
| XM_009292958.3 | XP_009291233.1 | 4 - 5 | 569 | 72 | ||
| XM_009292960.3 | XP_009291235.1 | 4 - 5 | 571 | 72 | ||
| XM_017351587.2 | XP_017207076.1 | 4 - 5 | 577 | 72 | ||
| GenBank mRNA | BC045890.1 | - | 4 - 5 | 477 | 72 | |
| BC153498.1 | - | 4 - 5 | 364 | 72 | ||
More Information
Set Membership: |
|

p4ha1b
Genomic Map