Assay Details
| Gene Symbol: p4ha1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_214691.1 | NP_999856.1 | 6 - 7 | 856 | 59 | |
| XM_009292958.3 | XP_009291233.1 | 6 - 7 | 948 | 59 | ||
| XM_009292960.3 | XP_009291235.1 | 6 - 7 | 950 | 59 | ||
| XM_017351587.2 | XP_017207076.1 | 6 - 7 | 956 | 59 | ||
| GenBank mRNA | BC045890.1 | - | 6 - 7 | 856 | 59 | |
| BC153498.1 | - | 6 - 7 | 743 | 59 | ||
More Information
Set Membership: |
|

p4ha1b
Genomic Map