Assay Details
| Gene Symbol: azi2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_214733.2 | NP_999898.2 | 4 - 5 | 581 | 56 | |
| GenBank mRNA | BC065984.1 | - | 4 - 5 | 548 | 56 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

azi2| Gene Symbol: azi2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_214733.2 | NP_999898.2 | 4 - 5 | 581 | 56 | |
| GenBank mRNA | BC065984.1 | - | 4 - 5 | 548 | 56 | |
Set Membership: |
|