Assay Details
| Gene Symbol: rbfox1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_214775.1 | NP_999940.1 | 1 - 2 | 85 | 84 | |
| GenBank mRNA | AB074763.1 | - | 1 - 2 | 85 | 84 | |
| BC163738.1 | - | 1 - 2 | 164 | 84 | ||
| BC163747.1 | - | 1 - 2 | 164 | 84 | ||
More Information
Set Membership: |
|

rbfox1l
Genomic Map