Assay Details
| Gene Symbol: rbfox1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_214775.1 | NP_999940.1 | 5 - 6 | 649 | 113 | |
| XM_005158173.4 | XP_005158230.1 | 5 - 6 | 701 | 113 | ||
| XM_021466671.1 | XP_021322346.1 | 5 - 6 | 710 | 113 | ||
| XM_021466672.1 | XP_021322347.1 | 6 - 7 | 683 | 113 | ||
| XM_021466673.1 | XP_021322348.1 | 6 - 7 | 932 | 113 | ||
| GenBank mRNA | AB074763.1 | - | 5 - 6 | 649 | 113 | |
| BC163738.1 | - | 5 - 6 | 728 | 113 | ||
| BC163747.1 | - | 5 - 6 | 728 | 113 | ||
More Information
Set Membership: |
|

rbfox1l
Genomic Map