Assay Details
| Gene Symbol: rabac1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_214780.1 | NP_999945.1 | 4 - 5 | 427 | 78 | |
| GenBank mRNA | AY391453.1 | - | 4 - 5 | 427 | 78 | |
| BC168498.1 | - | 4 - 5 | 450 | 78 | ||
More Information
Set Membership: |
|

rabac1
Genomic Map