Assay Details
| Gene Symbol: acta1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_214784.2 | NP_999949.2 | 1 - 2 | 46 | 62 | |
| GenBank mRNA | BC098546.1 | - | 1 - 2 | 46 | 62 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC098546.1, NM_214784.2 transcripts.
Set Membership: |
|

acta1b
Genomic Map