Assay Details
| Gene Symbol: asic1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_214790.2 | NP_999955.1 | 10 - 11 | 1475 | 83 | |
| XM_021478320.1 | XP_021333995.1 | 10 - 11 | 1733 | 83 | ||
| GenBank mRNA | AJ609616.1 | - | 10 - 11 | 1627 | 83 | |
| BC162330.1 | - | 10 - 11 | 1475 | 83 | ||
More Information
Set Membership: |
|

asic1a
Genomic Map