Assay Details
Gene Symbol: gpn2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_214801.2 | NP_999966.2 | 3 - 4 | 685 | 119 | |
XM_009292455.3 | XP_009290730.1 | 3 - 4 | 674 | 119 | ||
GenBank mRNA | AY576999.1 | - | 3 - 4 | 667 | 119 | |
BC083538.1 | - | 3 - 4 | 671 | 119 | ||
BC150450.1 | - | 3 - 4 | 662 | 119 | ||
BC152270.1 | - | 3 - 4 | 633 | 119 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
