Assay Details
| Gene Symbol: ppp1caa | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_214811.2 | NP_999976.1 | 5 - 6 | 1196 | 107 | |
| GenBank mRNA | BC070008.1 | - | 5 - 6 | 1149 | 107 | |
| BC154763.1 | - | 5 - 6 | 990 | 107 | ||
| CD758806.1 | - | 3 - 4 | 546 | 107 | ||
More Information
Set Membership: |
|

ppp1caa
Genomic Map