Assay Details
| Gene Symbol: adss1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_214820.1 | NP_999985.1 | 11 - 12 | 1184 | 82 | |
| GenBank mRNA | BC070009.1 | - | 11 - 12 | 1184 | 82 | |
More Information
Set Membership: |
|

adss1
Genomic Map