Assay Details
| Gene Symbol: adss1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_214820.1 | NP_999985.1 | 4 - 5 | 425 | 70 | |
| GenBank mRNA | BC070009.1 | - | 4 - 5 | 425 | 70 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

adss1| Gene Symbol: adss1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_214820.1 | NP_999985.1 | 4 - 5 | 425 | 70 | |
| GenBank mRNA | BC070009.1 | - | 4 - 5 | 425 | 70 | |
Set Membership: |
|