Assay Details
| Gene Symbol: pycr1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BC095354.1 | - | 2 - 3 | 199 | 119 | |
| BC164113.1 | - | 1 - 2 | 102 | 119 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

pycr1b| Gene Symbol: pycr1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BC095354.1 | - | 2 - 3 | 199 | 119 | |
| BC164113.1 | - | 1 - 2 | 102 | 119 | ||
Set Membership: |
|