Assay Details
| Gene Symbol: foxo3a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001009988.1 | NP_001009988.1 | 2 | 2348 | 80 | |
| GenBank mRNA | BC088386.1 | - | 2 | 2348 | 80 | |
More Information
Set Membership: |
|

foxo3a
Genomic Map