Assay Details
| Gene Symbol: rpe65b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001089433.2 | NP_001082902.2 | 14 | 1880 | 160 | |
| XM_021478150.1 | XP_021333825.1 | 11 | 1585 | 160 | ||
| GenBank mRNA | AY646887.1 | - | 14 | 1850 | 160 | |
More Information
Set Membership: |
|

rpe65b
Genomic Map