Assay Details
| Gene Symbol: kifc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001044954.2 | NP_001038419.1 | 17 | 2446 | 85 | |
| NM_131206.2 | NP_571281.2 | 17 | 2449 | 85 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

kifc1| Gene Symbol: kifc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001044954.2 | NP_001038419.1 | 17 | 2446 | 85 | |
| NM_131206.2 | NP_571281.2 | 17 | 2449 | 85 | ||
Set Membership: |
|