Assay Details
| Gene Symbol: actc1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_131591.1 | NP_571666.1 | 8 | 1150 | 151 | |
| GenBank mRNA | AF180887.1 | - | 8 | 1150 | 151 | |
| BC045406.1 | - | 9 | 1178 | 151 | ||
| BC065435.1 | - | 9 | 1158 | 151 | ||
| BC161649.1 | - | 2969 | 151 | |||
More Information
Set Membership: |
|

actc1b
Genomic Map