Assay Details
| Gene Symbol: LOC100147849 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021477626.1 | XP_021333301.1 | 36 | 9648 | 114 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100147849| Gene Symbol: LOC100147849 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021477626.1 | XP_021333301.1 | 36 | 9648 | 114 | |
Set Membership: |
|