Assay Details
| Gene Symbol: sypl2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_200667.2 | NP_956961.2 | 6 | 4035 | 117 | |
| GenBank mRNA | BC058043.1 | - | 7 | 3351 | 117 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

sypl2a| Gene Symbol: sypl2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_200667.2 | NP_956961.2 | 6 | 4035 | 117 | |
| GenBank mRNA | BC058043.1 | - | 7 | 3351 | 117 | |
Set Membership: |
|