Assay Details
| Gene Symbol: LOC100058651 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_023654514.1 | XP_023510282.1 | 1920 | 83 | ||
| XM_023654515.1 | XP_023510283.1 | 1943 | 83 | |||
More Information
Set Membership: |
|

LOC100058651
Genomic Map