Assay Details
| Gene Symbol: MMUT | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001309313.2 | NP_001296242.1 | 9 - 10 | 1853 | 79 | |
| XM_001502799.5 | XP_001502849.1 | 1951 | 79 | |||
| XM_014734210.2 | XP_014589696.1 | 1996 | 79 | |||
| XM_014734211.2 | XP_014589697.1 | 1960 | 79 | |||
| GenBank mRNA | HQ890001.1 | - | 4 - 5 | 427 | 79 | |
| JL640521.1 | - | 6 - 7 | 756 | 79 | ||
More Information
Set Membership: |
|

MMUT
Genomic Map