Assay Details
| Gene Symbol: PDIK1L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001504100.6 | XP_001504150.1 | 483 | 78 | ||
| XM_005607320.3 | XP_005607377.1 | 411 | 78 | |||
| XM_023635443.1 | XP_023491211.1 | 1021 | 78 | |||
| XM_023635444.1 | XP_023491212.1 | 428 | 78 | |||
More Information
Set Membership: |
|

PDIK1L
Genomic Map