Assay Details
| Gene Symbol: LOC100062210 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005602712.3 | XP_005602769.2 | 2954 | 109 | ||
| XM_023647955.1 | XP_023503723.1 | 2951 | 109 | |||
| XM_023647962.1 | XP_023503730.1 | 2798 | 109 | |||
More Information
Set Membership: |
|

LOC100062210
Genomic Map